Ethyl dodecanoate, 98%, Thermo Scientific Chemicals
Ethyl dodecanoate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl dodecanoate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
2500 mL
Catalog number A11188.AE
also known as A11188-AE
Price (USD)/ Each
40.65
Online exclusive
45.40 
Save 4.75 (10%)
-
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (USD)/ Each
40.65
Online exclusive
45.40 
Save 4.75 (10%)
Add to cart
Ethyl dodecanoate, 98%, Thermo Scientific Chemicals
Catalog numberA11188.AE
Price (USD)/ Each
40.65
Online exclusive
45.40 
Save 4.75 (10%)
-
Add to cart
Chemical Identifiers
CAS106-33-2
IUPAC Nameethyl dodecanoate
Molecular FormulaC14H28O2
InChI KeyMMXKVMNBHPAILY-UHFFFAOYSA-N
SMILESCCCCCCCCCCCC(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Identification (FTIR)Conforms
Appearance (Color)Clear colorless
Assay (GC)≥97.5%
Refractive Index1.4290-1.4330 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only