Thermo Scientific Chemicals

2-Mercapto-5-methylbenzimidazole, 98%, Thermo Scientific Chemicals

Catalog number: A11192.14
25 g, Each
Thermo Scientific Chemicals

2-Mercapto-5-methylbenzimidazole, 98%, Thermo Scientific Chemicals

Catalog number: A11192.14
25 g, Each
Quantity
Catalog number: A11192.14
also known as A11192-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
27231-36-3
IUPAC Name
5-methyl-2,3-dihydro-1H-1,3-benzodiazole-2-thione
Molecular Formula
C8H8N2S
InChI Key
CWIYBOJLSWJGKV-UHFFFAOYSA-N
SMILES
CC1=CC=C2NC(=S)NC2=C1
Form
Powder
Appearance (Color)
Pale cream to cream to pale brown
Assay (HPLC)
≥97.5%

Description

2-Mercapto-5-methylbenzimidazole is used as an additive in liquid paraffin. And also used in electrochemical techniques, corrosion inhibitors for steel under highly acidic conditions and pointed out that the effectiveness of the heterocyclic compound can be intimately associated to its molecular structure.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Mercapto-5-methylbenzimidazole is used as an additive in liquid paraffin. And also used in electrochemical techniques, corrosion inhibitors for steel under highly acidic conditions and pointed out that the effectiveness of the heterocyclic compound can be intimately associated to its molecular structure.

Solubility
Insoluble in water.

Notes
Hygroscopic. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text