Thermo Scientific Chemicals

4-Nitrophenyl isocyanate, 97%, Thermo Scientific Chemicals

Catalog number: A11194.06
5 g, Each
Thermo Scientific Chemicals

4-Nitrophenyl isocyanate, 97%, Thermo Scientific Chemicals

Catalog number: A11194.06
5 g, Each
Quantity
Catalog number: A11194.06
also known as A11194-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
100-28-7
IUPAC Name
1-isocyanato-4-nitrobenzene
Molecular Formula
C7H4N2O3
InChI Key
GFNKTLQTQSALEJ-UHFFFAOYSA-N
SMILES
[O-][N+](=O)C1=CC=C(C=C1)N=C=O
Identification (FTIR)
Conforms
Form
Crystals or powder or crystalline powder or fused solid
Melting Point (clear melt)
53.0-62.0?C
Appearance (Color)
Yellow
Assay (GC)
≥96.0%

Description

4-Nitrophenyl isocyanate is used as pharmaceutical intermediates. 4-Nitrophenyl isocyanate was used in the synthesis of 2-(5-methyl-3,4-diphenyl-1H-pyrrole-2-carbonyl)-N-(4-nitrophenyl)hydrazinecarboxamide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Nitrophenyl isocyanate is used as pharmaceutical intermediates. 4-Nitrophenyl isocyanate was used in the synthesis of 2-(5-methyl-3,4-diphenyl-1H-pyrrole-2-carbonyl)-N-(4-nitrophenyl)hydrazinecarboxamide.

Solubility
Insoluble in water.

Notes
Moisture Sensitive. Store at 2-8°C. Incompatible with water, heat and oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text