Thermo Scientific Chemicals

3,5-Dimethylbenzoic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A11198.09
10 g, Each
Thermo Scientific Chemicals

3,5-Dimethylbenzoic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A11198.09
10 g, Each
Quantity
Catalog number: A11198.09
also known as A11198-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
499-06-9
IUPAC Name
3,5-dimethylbenzoic acid
Molecular Formula
C9H10O2
InChI Key
UMVOQQDNEYOJOK-UHFFFAOYSA-N
SMILES
CC1=CC(=CC(C)=C1)C(O)=O
Melting Point (clear melt)
167.0-173.0?C
Appearance (Color)
Pale cream to cream to pale brown
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥98.0 to ≤102.0%
Assay (Silylated GC)
≥98.0%

Description

3,5-Dimethylbenzoic acid is suitable reagent used as carbon and energy supplement in the culture medium of Pseudomonas sp

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dimethylbenzoic acid is suitable reagent used as carbon and energy supplement in the culture medium of Pseudomonas sp

Solubility
Soluble in methanol. (1 g/10 mL). Slightly soluble in water.

Notes
Store away from oxidizing agents. Use only in a chemical fume hood. Incompatible with strong acids and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text