3,5-Dimethylbenzoic acid is suitable reagent used as carbon and energy supplement in the culture medium of Pseudomonas sp
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3,5-Dimethylbenzoic acid is suitable reagent used as carbon and energy supplement in the culture medium of Pseudomonas sp
Solubility
Soluble in methanol. (1 g/10 mL). Slightly soluble in water.
Notes
Store away from oxidizing agents. Use only in a chemical fume hood. Incompatible with strong acids and bases.
RUO – Research Use Only
Aliki Rontoyianni, Irene M. Mavridis. The crystal structure of the inclusion complex of cyclomaltoheptaose (β-cyclodextrin) with 3,5-dimethylbenzoic acid. Journal of Inclusion Phenomena and Macrocyclic Chemistry. 1994, 18 211-227.
J. M. Wilson; N. E. Gore; J. E. Sawbridge and F. Cardenas-Cruz. Acid-base equilibria of substituted benzoic acids. J. Chem. Soc. B. 1967, 852-859.