Thermo Scientific Chemicals

4'-Ethoxyacetanilide, 97%, Thermo Scientific Chemicals

Catalog number: A11200.36
500g, Each
Thermo Scientific Chemicals

4'-Ethoxyacetanilide, 97%, Thermo Scientific Chemicals

Catalog number: A11200.36
500g, Each
Quantity
Catalog number: A11200.36
also known as A11200-36
Price (USD)
Have Questions?

Chemical Identifiers

CAS
62-44-2
IUPAC Name
N-(4-ethoxyphenyl)acetamide
Molecular Formula
C10H13NO2
InChI Key
CPJSUEIXXCENMM-UHFFFAOYSA-N
SMILES
CCOC1=CC=C(NC(C)=O)C=C1
Appearance (Color)
White
Form
Crystals or powder or crystalline powder or flakes
Assay (GC)
≥96.0%
Identification (FTIR)
Conforms
Melting Point (clear melt)
131.0-140.0?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text