Thermo Scientific Chemicals

2-Carboxybenzaldehyde, 98+%, Thermo Scientific Chemicals

Catalog number: A11201.30
250 g, Each
Thermo Scientific Chemicals

2-Carboxybenzaldehyde, 98+%, Thermo Scientific Chemicals

Catalog number: A11201.30
250 g, Each
Quantity
Catalog number: A11201.30
also known as A11201-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
119-67-5
IUPAC Name
2-formylbenzoic acid
Molecular Formula
C8H6O3
InChI Key
DYNFCHNNOHNJFG-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=CC=C1C=O
Comment
Purchased in U.K. and U.S.
Appearance (Color)
White to cream
Assay (Aqueous acid-base Titration)
≥98.0 to ≤102.0% (U.K. specification)
Identification (FTIR)
Conforms (U.K. specification)
Assay from Supplier's CofA
≥98.0% (HPLC) (U.S. specification)

Description

2-Carboxybenzaldehyde is a carboxylated derivative of benzaldehyde which is readily converted to 2-hydroxymethyl benzoic acid by CBA dehydrogenase. 2-Carboxybenzaldehyde is a metabolite of ampicillin phthalidyl ester. It has been used to synthesize a series of N-substituted isoindolinones by reductive C-N coupling and intramolecular amidation with amines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Carboxybenzaldehyde is a carboxylated derivative of benzaldehyde which is readily converted to 2-hydroxymethyl benzoic acid by CBA dehydrogenase. 2-Carboxybenzaldehyde is a metabolite of ampicillin phthalidyl ester. It has been used to synthesize a series of N-substituted isoindolinones by reductive C-N coupling and intramolecular amidation with amines.

Solubility
Soluble in water.

Notes
Air Sensitive. Store away from air and oxidizing agents. Incompatible with strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text