Tris(3-chlorophenyl)phosphine, 98%, Thermo Scientific Chemicals
Tris(3-chlorophenyl)phosphine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tris(3-chlorophenyl)phosphine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number A11203.03
also known as A11203-03
Price (USD)/ Each
54.60
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
54.60
Add to cart
Tris(3-chlorophenyl)phosphine, 98%, Thermo Scientific Chemicals
Catalog numberA11203.03
Price (USD)/ Each
54.60
-
Add to cart
Chemical Identifiers
CAS29949-85-7
IUPAC Nametris(3-chlorophenyl)phosphane
Molecular FormulaC18H12Cl3P
InChI KeyQAPGHLJQIVDTPT-UHFFFAOYSA-N
SMILESClC1=CC(=CC=C1)P(C1=CC(Cl)=CC=C1)C1=CC=CC(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Melting Point (clear melt)60.0-67.0?C
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only