2,3-Dichlorotoluene, 98%, Thermo Scientific Chemicals
2,3-Dichlorotoluene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,3-Dichlorotoluene, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
50 g
100 g
Catalog number A11206.14
also known as A11206-14
Price (USD)/ Each
65.20
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
65.20
Add to cart
2,3-Dichlorotoluene, 98%, Thermo Scientific Chemicals
Catalog numberA11206.14
Price (USD)/ Each
65.20
-
Add to cart
Chemical Identifiers
CAS32768-54-0
IUPAC Name1,2-dichloro-3-methylbenzene
Molecular FormulaC7H6Cl2
InChI KeyGWLKCPXYBLCEKC-UHFFFAOYSA-N
SMILESCC1=CC=CC(Cl)=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Identification (FTIR)Conforms
Refractive Index1.5495-1.5535 @ 20?C
FormLiquid
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only