Thermo Scientific Chemicals

4'-Chloroacetanilide, 98+%, Thermo Scientific Chemicals

Catalog number: A11214.14
25 g, Each
Thermo Scientific Chemicals

4'-Chloroacetanilide, 98+%, Thermo Scientific Chemicals

Catalog number: A11214.14
25 g, Each
Quantity
Catalog number: A11214.14
also known as A11214-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
539-03-7
IUPAC Name
N-(4-chlorophenyl)acetamide
Molecular Formula
C8H8ClNO
InChI Key
GGUOCFNAWIODMF-UHFFFAOYSA-N
SMILES
CC(=O)NC1=CC=C(Cl)C=C1
Identification (FTIR)
Conforms
Melting Point (clear melt)
177-182?C
Appearance (Color)
White to cream to pale brown or grey
Form
Crystals or powder or crystalline powder
Assay (GC)
≥98.0%

Description

4'-Chloroacetanilide is used in pharmaceutical preparations, using PGC column. And also used in the synthesis of alprazolam.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′-Chloroacetanilide is used in pharmaceutical preparations, using PGC column. And also used in the synthesis of alprazolam.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text