Thermo Scientific Chemicals

Ethyl difluoroacetate, 98%, Thermo Scientific Chemicals

Catalog number: A11215.06
5 g, Each
Thermo Scientific Chemicals

Ethyl difluoroacetate, 98%, Thermo Scientific Chemicals

Catalog number: A11215.06
5 g, Each
Quantity
Catalog number: A11215.06
also known as A11215-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
454-31-9
IUPAC Name
ethyl 2,2-difluoroacetate
Molecular Formula
C4H6F2O2
InChI Key
GZKHDVAKKLTJPO-UHFFFAOYSA-N
SMILES
CCOC(=O)C(F)F
Appearance (Color)
Clear colorless
Assay (GC)
≥97.5%
Refractive Index
1.3455-1.3495 @ 20?C
Form
Liquid

Description

Ethyl difluoroacetate is used in pharmaceutical intermediates, acrylization oxidation and halogenating reaction of catalyst. Also ethyl difluoroacetate is used in preparing pyrrazolo-pyrimidine derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl difluoroacetate is used in pharmaceutical intermediates, acrylization oxidation and halogenating reaction of catalyst. Also ethyl difluoroacetate is used in preparing pyrrazolo-pyrimidine derivatives.

Solubility
Slightly soluble in water

Notes
Hygroscopic. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text