Thermo Scientific Chemicals

Sodium hexanitritocobaltate(III), Thermo Scientific Chemicals

Catalog number: A11216.30
250 g, Each
Thermo Scientific Chemicals

Sodium hexanitritocobaltate(III), Thermo Scientific Chemicals

Catalog number: A11216.30
250 g, Each
Quantity
Catalog number: A11216.30
also known as A11216-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14649-73-1
IUPAC Name
cobalt(3+) trisodium hexakis(dioxoamine)
Molecular Formula
CoH6N6Na3O12
InChI Key
QLNJFTGEDYRKNP-UHFFFAOYSA-N
SMILES
[Na+].[Na+].[Na+].[Co+3].[O-][NH+]=O.[O-][NH+]=O.[O-][NH+]=O.[O-][NH+]=O.[O-][NH+]=O.[O-][NH+]=O
Form
Crystalline powder
Comment
Accepted on Supplier's Certificate of Analysis
Comment
Suitable for the determination of potassium
Insoluble material
≤0.02%
Appearance (Color)
Yellow to Orange

Description

Sodium hexanitritocobaltate(III) is used as primary and secondary intermediate in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium hexanitritocobaltate(III) is used as primary and secondary intermediate in chemical research.

Solubility
Soluble in water.

Notes
Store at room temperature. Incompatible with acids, reducing agents and amines.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text