Thermo Scientific Chemicals

2-Methylcyclohexane-1,3-dione, 98+%, Thermo Scientific Chemicals

Catalog number: A11218.06
5 g, Each
Thermo Scientific Chemicals

2-Methylcyclohexane-1,3-dione, 98+%, Thermo Scientific Chemicals

Catalog number: A11218.06
5 g, Each
Quantity
Catalog number: A11218.06
also known as A11218-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1193-55-1
Assay (HPLC)
≥98.0%
Identification (FTIR)
Conforms
Loss on Drying
≤3% (105°C/constant weight)
Appearance (Color)
White to cream to pale yellow
Form
Crystals or powder or crystalline powder

Description

2-Methylcyclohexane-1,3-dione acts as a synthetic intermediate. It is an important starting material, exemplified in a formal synthesis of acorone. It is also used in Organic Synthesis, Pharmaceuticals, Agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylcyclohexane-1,3-dione acts as a synthetic intermediate. It is an important starting material, exemplified in a formal synthesis of acorone. It is also used in Organic Synthesis, Pharmaceuticals, Agrochemicals.

Solubility
Soluble in methanol ( 0.1 g/mL). Insoluble in water.

Notes
Hygroscopic. Store at -20°C. Incompatible with oxidizing agents and moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text