Thermo Scientific Chemicals

5-Bromo-1-pentene, 96%, Thermo Scientific Chemicals

Catalog number: A11219.06
5 g, Each
Thermo Scientific Chemicals

5-Bromo-1-pentene, 96%, Thermo Scientific Chemicals

Catalog number: A11219.06
5 g, Each
Quantity
Catalog number: A11219.06
also known as A11219-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1119-51-3
IUPAC Name
5-bromopent-1-ene
Molecular Formula
C5H9Br
InChI Key
LPNANKDXVBMDKE-UHFFFAOYSA-N
SMILES
BrCCCC=C
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥95.0% (UK-sourced material)
Comment
Material sourced in UK and US
Form
Liquid
Assay from Supplier's CofA
≥95.0% (US-sourced material)

Description

5-Bromo-1-pentene is widely utilized in the stereoselective synthesis of 7alpha-(3-carboxypropyl) estradiol. It plays an important role as starting material in the synthesis of a variety of compounds including DL-histrionicotoxin and benzophenone containing fatty acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromo-1-pentene is widely utilized in the stereoselective synthesis of 7alpha-(3-carboxypropyl) estradiol. It plays an important role as starting material in the synthesis of a variety of compounds including DL-histrionicotoxin and benzophenone containing fatty acids.

Solubility
Immiscible with water.

Notes
Keep away from sources of ignition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text