3,4-(Methylenedioxy)phenylacetic acid, 98%, Thermo Scientific Chemicals
3,4-(Methylenedioxy)phenylacetic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,4-(Methylenedioxy)phenylacetic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Catalog number A11229.14
also known as A11229-14
Price (USD)/ Each
286.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
286.00
Add to cart
3,4-(Methylenedioxy)phenylacetic acid, 98%, Thermo Scientific Chemicals
Catalog numberA11229.14
Price (USD)/ Each
286.00
-
Add to cart
Chemical Identifiers
CAS2861-28-1
IUPAC Name2-(2H-1,3-benzodioxol-5-yl)acetic acid
Molecular FormulaC9H8O4
InChI KeyODVLMCWNGKLROU-UHFFFAOYSA-N
SMILESOC(=O)CC1=CC=C2OCOC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥97.5%
Appearance (Color)White to cream to pale brown
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Melting Point (clear melt)127.0-133.0?C
FormCrystals or powder or crystalline powder
3,4-(Methylenedioxy)phenylacetic acid is used as organic intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-(Methylenedioxy)phenylacetic acid is used as organic intermediates.

Solubility
Soluble in water (partly), ethanol, and methanol.

Notes
Store at room temperature. Store away from oxidizing agents. Store in a cool, dry place. Store in a tightly closed container.
RUO – Research Use Only
Carlos Camacho Camacho; Dick de Vos; Bernard Mahieu; Marcel Gielen; Martine Kemmer; Monique Biesemans; Rudolph Willem. Organotin(Iv) Derivatives Of 3,4-(Methylenedioxy) Phenylacetic Acid: Synthesis, Spectroscopic Characterization And In Vitro Antitumour Properties.Main Group Metal Chemistry. 2000, 23 381-386.