Thermo Scientific Chemicals

4-Acetamidophenol, 98%, Thermo Scientific Chemicals

Catalog number: A11240.0I
5000 g, Each
Thermo Scientific Chemicals

4-Acetamidophenol, 98%, Thermo Scientific Chemicals

Catalog number: A11240.0I
5000 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A11240.0I
also known as A11240-0I
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
103-90-2
IUPAC Name
N-(4-hydroxyphenyl)acetamide
Molecular Formula
C8H9NO2
InChI Key
RZVAJINKPMORJF-UHFFFAOYSA-N
SMILES
CC(=O)NC1=CC=C(O)C=C1
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
167.0-173.0?C
Appearance (Color)
White
Assay (HPLC)
≥97.5%
Identification (FTIR)
Conforms

Description

Cyclooxygenase inhibitor and widely used analgesic. 4-Acetamidophenol formulated pharmaceutical products are used as antiinfectant, analgesic, anti rheumatic and antipyretic. It is used as an intermediate in organic synthesis, hydrogen peroxide stabilizer and photographic chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclooxygenase inhibitor and widely used analgesic. 4-Acetamidophenol formulated pharmaceutical products are used as antiinfectant, analgesic, anti rheumatic and antipyretic. It is used as an intermediate in organic synthesis, hydrogen peroxide stabilizer and photographic chemicals.

Solubility
Soluble in alcohol, dimethylformamide, ethylene dichloride, acetone and ethyl acetate. Slightly soluble in water and ether.

Notes
Incompatible with strong oxidizing agents, acids and alkali metals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text