4-Chromanone has been used to study the substrate specificity and catalytic ability of 4-hydroxyacetophenone monooxygenase isolated from Pseudomonas fluorescens ACB
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Chromanone has been used to study the substrate specificity and catalytic ability of 4-hydroxyacetophenone monooxygenase isolated from Pseudomonas fluorescens ACB
Solubility
Slightly soluble in water.
Notes
Incompatible with oxidizing agents and moisture. Store away from oxidizing agents. Store at 2-8°C.
RUO – Research Use Only
Yao-Jun Shi; Dongwei Cai; U.-H. Dolling; Alan W. Douglas; David M. Tschaen; Thomas R. Verhoeven. An improved method for chiral oxazaborolidine-catalyzed reduction of 4-chromanone analogs and MK-0499. Tetrahedron letters 1994, 35, 6409-6412.