Sodium perchlorate monohydrate, 97+%, Thermo Scientific Chemicals
Sodium perchlorate monohydrate, 97+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium perchlorate monohydrate, 97+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1000 g
250 g
5000 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A11243.0B
also known as A11243-0B
Price (USD)/ Each
153.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
180.00 
Save 26.35 (15%)
-
Add to cart
Quantity:
1000 g
Request bulk or custom format
Price (USD)/ Each
153.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
180.00 
Save 26.35 (15%)
Add to cart
Sodium perchlorate monohydrate, 97+%, Thermo Scientific Chemicals
Catalog numberA11243.0B
Price (USD)/ Each
153.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
180.00 
Save 26.35 (15%)
-
Add to cart
Chemical Identifiers
CAS7791-07-3
IUPAC Namesodium hydrate perchlorate
Molecular FormulaClH2NaO5
InChI KeyIXGNPUSUVRTQGW-UHFFFAOYSA-M
SMILESO.[Na+].[O-][Cl](=O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormAdhering crystals (U.K. specification)
FormCrystalline powder or crystals or chunks and/or compacted solid (non-U.K. specification)
Assay (special)≥97.0% (non-U.K. specification)
Insoluble material≤0.005% (U.K. specification)
Water Content (Karl Fischer Titration)7.5-15.0% (non-U.K. specification)
View more
Sodium perchlorate, monohydrate is a chaotropic agent used in standard DNA extraction and hybridization reactions in molecular biology. It is widely utilized as a deproteinization agent during nucleic acid synthesis. A bit of modern commercial explosives products may contain sodium perchlorate, ammonium perchlorate, or potassium perchlorate as a chemical sensitizing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium perchlorate, monohydrate is a chaotropic agent used in standard DNA extraction and hybridization reactions in molecular biology. It is widely utilized as a deproteinization agent during nucleic acid synthesis. A bit of modern commercial explosives products may contain sodium perchlorate, ammonium perchlorate, or potassium perchlorate as a chemical sensitizing agent.

Solubility
Soluble in water.

Notes
It is a hygroscopic solid. Store under inert atmosphere and keep away from ignition sources.
RUO – Research Use Only

General References:

  1. Ghosh, K.; Mohan, V.; Kumar, P.;. Ng, S. W.; Tiekin, E. R. T. Selective fluorescence sensing of Ni2+ by tetradentate ligands: Synthesis of nickel complexes and crystal structures. Inorg. Chim. Acta. 2014, 416, 76-84.
  2. Polonyi, C.; Alshiekh, A.; Sarsam, L. A.; Clausén, M.; Elmroth, S. K. C. Cisplatin-induced duplex dissociation of complementary and destabilized short GG-containing duplex RNAs. Dalton Trans. 2014, 43, 11941-11949.