Thermo Scientific Chemicals

Sodium perchlorate monohydrate, 97+%, Thermo Scientific Chemicals

Catalog number: A11243.30
250 g, Each
Thermo Scientific Chemicals

Sodium perchlorate monohydrate, 97+%, Thermo Scientific Chemicals

Catalog number: A11243.30
250 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A11243.30
also known as A11243-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7791-07-3
IUPAC Name
sodium hydrate perchlorate
Molecular Formula
ClH2NaO5
InChI Key
IXGNPUSUVRTQGW-UHFFFAOYSA-M
SMILES
O.[Na+].[O-][Cl](=O)(=O)=O
Form
Adhering crystals (U.K. specification)
Form
Crystalline powder or crystals or chunks and/or compacted solid (non-U.K. specification)
Assay (special)
≥97.0% (non-U.K. specification)
Insoluble material
≤0.005% (U.K. specification)
Water Content (Karl Fischer Titration)
7.5-15.0% (non-U.K. specification)

Description

Sodium perchlorate, monohydrate is a chaotropic agent used in standard DNA extraction and hybridization reactions in molecular biology. It is widely utilized as a deproteinization agent during nucleic acid synthesis. A bit of modern commercial explosives products may contain sodium perchlorate, ammonium perchlorate, or potassium perchlorate as a chemical sensitizing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium perchlorate, monohydrate is a chaotropic agent used in standard DNA extraction and hybridization reactions in molecular biology. It is widely utilized as a deproteinization agent during nucleic acid synthesis. A bit of modern commercial explosives products may contain sodium perchlorate, ammonium perchlorate, or potassium perchlorate as a chemical sensitizing agent.

Solubility
Soluble in water.

Notes
It is a hygroscopic solid. Store under inert atmosphere and keep away from ignition sources.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text