Thermo Scientific Chemicals

Undecanoic acid, 98%, Thermo Scientific Chemicals

Catalog number: A11244.18
50 g, Each
Thermo Scientific Chemicals

Undecanoic acid, 98%, Thermo Scientific Chemicals

Catalog number: A11244.18
50 g, Each
Quantity
Catalog number: A11244.18
also known as A11244-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
112-37-8
IUPAC Name
undecanoic acid
Molecular Formula
C11H22O2
InChI Key
ZDPHROOEEOARMN-UHFFFAOYSA-N
SMILES
CCCCCCCCCCC(O)=O
Assay (Silylated GC)
> 97.5%
Form
fused solid, clear colourless as melt
Assay (Aqueous acid-base Titration)
> 97.5%
Appearance (Color)
White or translucent

Description

Undecanoic acid is used as an unsaturated fatty acid. It is also used as intermediates of Liquid Crystals. It is used in organic synthesis, Catalytic agent, Petrochemical additive. It is also used in anti-fungal agent, perfumery chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Undecanoic acid is used as an unsaturated fatty acid. It is also used as intermediates of Liquid Crystals. It is used in organic synthesis, Catalytic agent, Petrochemical additive. It is also used in anti-fungal agent, perfumery chemicals.

Solubility
Insoluble in water.

Notes
Store at 4°C. Incompatible with bases, oxidizing agents, reducing agents. Keep container closed when not in use.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text