Phenylhydrazine, 97%, Thermo Scientific Chemicals
Phenylhydrazine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Phenylhydrazine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Catalog number A11246.30
also known as A11246-30
Price (USD)/ Each
69.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
81.40 
Save 11.75 (14%)
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
69.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
81.40 
Save 11.75 (14%)
Add to cart
Phenylhydrazine, 97%, Thermo Scientific Chemicals
Catalog numberA11246.30
Price (USD)/ Each
69.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
81.40 
Save 11.75 (14%)
-
Add to cart
Chemical Identifiers
CAS100-63-0
IUPAC Namehydrogen phenylhydrazine chloride
Molecular FormulaC6H9ClN2
InChI KeyJOVOSQBPPZZESK-UHFFFAOYSA-N
SMILES[H+].[Cl-].NNC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to orange to brown
Assay (Non-aqueous acid-base Titration)≥96.0% (UK-sourced material)
CommentMaterial sourced in UK and US
FormLiquid or fused solid
Assay from Supplier's CofA≥96.0% (US-sourced material)
View more
Phenyl hydrazine is involved in Fischer indole synthesis to prepare indoles, which find application as intermediates in the pharmaceuticals particularly for the tryptamine drug. In analytical chemistry, it is used to differentiate and separate sugars by forming phenyhydrazones. It is used as an N-protecting reagent and for cleavage of phthaloyl group.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenyl hydrazine is involved in Fischer indole synthesis to prepare indoles, which find application as intermediates in the pharmaceuticals particularly for the tryptamine drug. In analytical chemistry, it is used to differentiate and separate sugars by forming phenyhydrazones. It is used as an N-protecting reagent and for cleavage of phthaloyl group.

Solubility
Soluble in dilute acids.

Notes
Air and light sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

General References:

  1. Derivatizing agent for the conversion of carbonyl compounds to their phenylhydrazones best known as intermediates in the Fischer indole synthesis, by cyclization under acidic conditions with loss of ammonia. The classical reagent is anhydrous ZnCl2: Org. Synth. Coll., 3, 725 (1976). For reviews, see: Chem. Rev., 63, 373 (1963); 69, 227 (1969); Tetrahedron, 36, 161 (1980); Org. Prep. Proced. Int., 25, 609 (1993). Milder cyclization conditions are sometimes effective, e.g.: dilute H2SO4: J. Org. Chem., 59, 3738 (1994); PCl3: J. Chem. Soc., Chem. Commun., 563 (1981); Tetrahedron, 41, 4615 (1985). Cyclohexanone phenylhydrazone gives tetrahydrocarbazole on heating in acetic acid: Org. Synth. Coll., 4, 884 (1963).
  2. Methods used for the cleavage of phenylhydrazones include exchange by refluxing with acetone: J. Org. Chem., 40, 3302 (1975), or oxidative cleavage by Fe(NO3)3 on a clay support: Synthesis, 439 (1985).
  3. Wang, L.; Wang, P.; Liu, Y.; Zheng, L.; Sun, Q.; Qiu, S.; Liu, C. Effects of phenylhydrazine-4-sulfonic acid on the reduction of GO and preparation of hydrophilic graphene with broad pH stability and antioxidant activity. RSC Adv. 2015, 5 (48), 38696-38705.
  4. Gein, V. L.; Mar’yasov, M. A. Reactions of 5-aryl-4-(hetaren-2-ylcarbonyl)-3-hydroxy-1-(1, 3-thiazol-2-yl)-2, 5-dihydro-1H-pyrrol-2-ones with hydrazine, phenylhydrazine, and hydroxylamine. Russ. J. Org. Chem. 2015, 51 (1), 110-115.