2-Phenoxynicotinic acid, 98%, Thermo Scientific Chemicals
2-Phenoxynicotinic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Phenoxynicotinic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number A11247.03
also known as A11247-03
Price (USD)/ Each
28.30
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
28.30
Add to cart
2-Phenoxynicotinic acid, 98%, Thermo Scientific Chemicals
Catalog numberA11247.03
Price (USD)/ Each
28.30
-
Add to cart
Chemical Identifiers
CAS35620-71-4
IUPAC Name2-phenoxypyridine-3-carboxylate
Molecular FormulaC12H8NO3
InChI KeyCQGAXJGXGLVFGJ-UHFFFAOYSA-M
SMILES[O-]C(=O)C1=CC=CN=C1OC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream to cream
FormPowder
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (Silylated GC)≥97.5%
Melting Point (clear melt)177.0-184.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only