Thermo Scientific Chemicals

D-(-)-Tartaric acid, 99%, Thermo Scientific Chemicals

Catalog number: A11264.22
100 g, Each
Thermo Scientific Chemicals

D-(-)-Tartaric acid, 99%, Thermo Scientific Chemicals

Catalog number: A11264.22
100 g, Each
Quantity
Catalog number: A11264.22
also known as A11264-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
147-71-7
IUPAC Name
2,3-dihydroxybutanedioic acid
Molecular Formula
C4H6O6
InChI Key
FEWJPZIEWOKRBE-UHFFFAOYNA-N
SMILES
OC(C(O)C(O)=O)C(O)=O
Melting Point (clear melt)
163.0-172.0?C (UK-sourced material)
Optical Rotation
-13.5 ± 0.5? (C=10 in water, UK-sourced material)
Appearance (Color)
White
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5% (UK-sourced material)

Description

D-(-)-Tartaric acid is used as a resolving agent in organic synthesis. It is used as a precursor for the preparation of its ester derivatives like D-tartaric acid diethyl ester, D-tartaric acid dimethyl ester and D-tartaric acid diiso-propyl ester. It finds application in the synthesis of chiral aziridine derivative, a common intermediate for the preparation of hydroxyethylamine class HIV protease inhibitors viz. as saquinavir, amprenavir and nelfinavir. It is widely used in the food industry as a beer foaming agent, for food acidity regulations and as a flavoring agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-(-)-Tartaric acid is used as a resolving agent in organic synthesis. It is used as a precursor for the preparation of its ester derivatives like D-tartaric acid diethyl ester, D-tartaric acid dimethyl ester and D-tartaric acid diiso-propyl ester. It finds application in the synthesis of chiral aziridine derivative, a common intermediate for the preparation of hydroxyethylamine class HIV protease inhibitors viz. as saquinavir, amprenavir and nelfinavir. It is widely used in the food industry as a beer foaming agent, for food acidity regulations and as a flavoring agent.

Solubility
Soluble in water and alcohol.

Notes
Light sensitive. Incompatible with bases, oxidizing agents and reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text