Sodium fumarate, 98%, Thermo Scientific Chemicals
Sodium fumarate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium fumarate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2500 g
Catalog number A11276.36
also known as A11276-36
Price (USD)/ Each
107.65
Online exclusive
119.00 
Save 11.35 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
107.65
Online exclusive
119.00 
Save 11.35 (10%)
Add to cart
Sodium fumarate, 98%, Thermo Scientific Chemicals
Catalog numberA11276.36
Price (USD)/ Each
107.65
Online exclusive
119.00 
Save 11.35 (10%)
-
Add to cart
Chemical Identifiers
CAS17013-01-3
IUPAC Namedisodium (2Z)-but-2-enedioate
Molecular FormulaC4H2Na2O4
InChI KeyMSJMDZAOKORVFC-UAIGNFCESA-L
SMILES[Na+].[Na+].[O-]C(=O)\C=C/C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5%
Water Content (Karl Fischer Titration)≤1.0%
Appearance (Color)White
Identification (FTIR)Conforms
Sodium fumarate is used as an acidity regulator in food processing and as terminal electron acceptors in the cultivation of certain anaerobic microorganisms. It is also used as a buffering agent and pH adjuster in cosmetic and personal care products. Further, it is used as a food additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium fumarate is used as an acidity regulator in food processing and as terminal electron acceptors in the cultivation of certain anaerobic microorganisms. It is also used as a buffering agent and pH adjuster in cosmetic and personal care products. Further, it is used as a food additive.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only

General References:

  1. Harry-O’kuru, R. E.; Gordon, S. H.; Klokkenga, M. Bio-generation of succinic acid by fermentation of Physaria fendleri seed polysaccharides. Ind. Crops Prod. 2015, 77, 116-122.
  2. Staszak, K.; Woźniak, M.; Sottek, M.; Karaś, Z.; Prochaska, K. Removal of fumaric acid from simulated and real fermentation broth. J. Chem. Technol. Biotechnol. 2015, 90 (3), 432-440.