Thermo Scientific Chemicals

(3-Aminopropyl)trimethoxysilane, 97%, Thermo Scientific Chemicals

Catalog number: A11284.0E
2500 g, Each
Thermo Scientific Chemicals

(3-Aminopropyl)trimethoxysilane, 97%, Thermo Scientific Chemicals

Catalog number: A11284.0E
2500 g, Each
Quantity
Catalog number: A11284.0E
also known as A11284-0E
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13822-56-5
IUPAC Name
(3-aminopropyl)trimethoxysilane
Molecular Formula
C6H17NO3Si
InChI Key
SJECZPVISLOESU-UHFFFAOYSA-N
SMILES
CO[Si](CCCN)(OC)OC
Appearance (Color)
Clear colorless
Refractive Index
1.4205-1.4255 @ 20?C
Assay (GC)
≥96.0%
Form
Liquid
Identification (FTIR)
Conforms

Description

(3-Aminopropyl)trimethoxysilane is used as a silane coupling agent on silver nanoparticles. It is also used in silanization processes. It is also used to prepare surface modified titanate nanotubes (TiNT).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(3-Aminopropyl)trimethoxysilane is used as a silane coupling agent on silver nanoparticles. It is also used in silanization processes. It is also used to prepare surface modified titanate nanotubes (TiNT).

Solubility
Miscible with water.

Notes
Moisture sensitive. Incompatible withstrong acids, strong oxidizing agents, water, bases, alcohols and peroxides.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text