2,6-Difluorobenzoyl chloride, 98%, Thermo Scientific Chemicals
2,6-Difluorobenzoyl chloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,6-Difluorobenzoyl chloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A11286.06
also known as A11286-06
Price (USD)/ Each
29.50
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
29.50
Add to cart
2,6-Difluorobenzoyl chloride, 98%, Thermo Scientific Chemicals
Catalog numberA11286.06
Price (USD)/ Each
29.50
-
Add to cart
Chemical Identifiers
CAS18063-02-0
IUPAC Name2,6-difluorobenzoyl chloride
Molecular FormulaC7H3ClF2O
InChI KeyQRHUZEVERIHEPT-UHFFFAOYSA-N
SMILESFC1=CC=CC(F)=C1C(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Refractive Index1.4970-1.5015 @ 20?C
FormLiquid
Appearance (Color)Clear colorless to pale yellow
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only