Thermo Scientific Chemicals

1,1-Diphenylhydrazine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: A11289.22
100 g, Each
Thermo Scientific Chemicals

1,1-Diphenylhydrazine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: A11289.22
100 g, Each
Quantity
Catalog number: A11289.22
also known as A11289-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
530-47-2
IUPAC Name
hydrogen 1,1-diphenylhydrazine chloride
Molecular Formula
C12H13ClN2
InChI Key
MIVUDWFNUOXEJM-UHFFFAOYSA-N
SMILES
[H+].[Cl-].NN(C1=CC=CC=C1)C1=CC=CC=C1
Assay (Titration ex Chloride)
≥97.5 to ≤102.5%
Form
Powder
Assay (Freebase GC)
≥97.5%
Appearance (Color)
to pale gray to gray

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text