Potassium perchlorate, 99%, Thermo Scientific Chemicals
Potassium perchlorate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium perchlorate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A11296.36
Price (USD)/ Each
101.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
119.00 
Save 17.35 (15%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
101.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
119.00 
Save 17.35 (15%)
Add to cart
Potassium perchlorate, 99%, Thermo Scientific Chemicals
Catalog numberA11296.36
Price (USD)/ Each
101.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
119.00 
Save 17.35 (15%)
-
Add to cart
Chemical Identifiers
CAS7778-74-7
IUPAC Namepotassium perchlorate
Molecular FormulaClKO4
InChI KeyYLMGFJXSLBMXHK-UHFFFAOYSA-M
SMILES[K+].[O-][Cl](=O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Purity (DSC)≥98.5% (non-U.S. sourced material)
Assay from Suppliers CofA≥98.5% (U.S. sourced material)
Melting Point (DSC)299.0-306.0°C (non-U.S. sourced material)
Potassium perchlorate is used in ammunition percussion caps, explosive primers, fireworks and propellants. It acts as a solid rocket propellant. It is also used as an oxidizer in colored pyrotechnic compositions and flash powder. It is used as an antithyroid and involved in the treatment of hyperthyroidism. The mixture of anthracene and sulfur with potassium perchlorate is used for generating black smoke signaling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium perchlorate is used in ammunition percussion caps, explosive primers, fireworks and propellants. It acts as a solid rocket propellant. It is also used as an oxidizer in colored pyrotechnic compositions and flash powder. It is used as an antithyroid and involved in the treatment of hyperthyroidism. The mixture of anthracene and sulfur with potassium perchlorate is used for generating black smoke signaling.

Solubility
Soluble in water. Slightly soluble in acetone. Insoluble in ethanol, ethyl acetate and ether.

Notes
Hygroscopic. Incompatible with strong reducing agents, powdered metals, strong acids and alcohols.
RUO – Research Use Only

General References:

  1. Pourmortazavi, S. M.; Zaree, A.; Mirsadeghi, S. Taguchi approach-assisted optimization of spraying in non-solvent process for preparation of potassium perchlorate nanoparticles. J. Sol-Gel Sci. Technol. 2015, 76 (3), 510-518.
  2. Dolgoborodov, A. Y.; Ermolaev, B. S.; Shevchenko, A. A.; Teselkin, V. A.; Kirilenko, V. G.; Monogarov, K. A.; Streletskii, A. N. Combustion and detonation of mechanoactivated aluminum-potassium perchlorate mixtures. Russ. J. Phys. Chem. B 2015, 9 (4), 615-624.