Thermo Scientific Chemicals

Tropolone, 98%, Thermo Scientific Chemicals

Catalog number: A11299.03
1 g, Each
Thermo Scientific Chemicals

Tropolone, 98%, Thermo Scientific Chemicals

Catalog number: A11299.03
1 g, Each
Quantity
Catalog number: A11299.03
also known as A11299-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
533-75-5
IUPAC Name
2-hydroxycyclohepta-2,4,6-trien-1-one
Molecular Formula
C7H6O2
InChI Key
MDYOLVRUBBJPFM-UHFFFAOYSA-N
SMILES
OC1=CC=CC=CC1=O
Appearance (Color)
White to cream to pale brown or pale yellow
Form
Crystals or powder or crystalline powder
Assay (GC)
≥97.5%
Water Content (Karl Fischer Titration)
<1.0%
Melting Point (clear melt)
50-55?C

Description

Tropolone is a sensitive reagent for reducing sugars. A non-benzenoid aromatic compound, Reagent for the preparation of fused heterocycles and complexes of Ga(III) and In(III).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text