Thermo Scientific Chemicals

Barium nitrate, 99%, Thermo Scientific Chemicals

Catalog number: A11305.36
500 g, Each
Thermo Scientific Chemicals

Barium nitrate, 99%, Thermo Scientific Chemicals

Catalog number: A11305.36
500 g, Each
Quantity
Catalog number: A11305.36
also known as A11305-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10022-31-8
IUPAC Name
barium(2+) dinitrate
Molecular Formula
BaN2O6
InChI Key
IWOUKMZUPDVPGQ-UHFFFAOYSA-N
SMILES
[Ba++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms
Appearance (Color)
White to pale cream
Complexometric Titration
≥98.5 to ≤101.5%

Description

Barium nitrate is used in the manufacture of special glasses, in fire works industries and for green-fire in pyrotechnics. It is widely used in defense applications. It is a precursor material for barium oxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Barium nitrate is used in the manufacture of special glasses, in fire works industries and for green-fire in pyrotechnics. It is widely used in defense applications. It is a precursor material for barium oxide.

Solubility
Soluble in water. Insoluble in alcohol.

Notes
Moisture sensitive. Incompatible with metal,alloy powders, combustible materials, reducing agents, acids, and acid anhydrides.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text