2,5-Dichlorobenzotrifluoride, 98%, Thermo Scientific Chemicals
2,5-Dichlorobenzotrifluoride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,5-Dichlorobenzotrifluoride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number A11310.14
also known as A11310-14
Price (USD)/ Each
78.70
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
78.70
Add to cart
2,5-Dichlorobenzotrifluoride, 98%, Thermo Scientific Chemicals
Catalog numberA11310.14
Price (USD)/ Each
78.70
-
Add to cart
Chemical Identifiers
CAS320-50-3
IUPAC Name1,4-dichloro-2-(trifluoromethyl)benzene
Molecular FormulaC7H3Cl2F3
InChI KeyDYBYUWVMLBBEMA-UHFFFAOYSA-N
SMILESFC(F)(F)C1=CC(Cl)=CC=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.4805-1.4845 @ 20?C
Assay (GC)≥97.5%
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only