2,2,2-Trimethylacetamide, 98+%, Thermo Scientific Chemicals
2,2,2-Trimethylacetamide, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,2,2-Trimethylacetamide, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
250 g
Catalog number A11319.18
also known as A11319-18
Price (USD)/ Each
153.00
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
153.00
Add to cart
2,2,2-Trimethylacetamide, 98+%, Thermo Scientific Chemicals
Catalog numberA11319.18
Price (USD)/ Each
153.00
-
Add to cart
Chemical Identifiers
CAS754-10-9
IUPAC Name2,2-dimethylpropanamide
Molecular FormulaC5H11NO
InChI KeyXIPFMBOWZXULIA-UHFFFAOYSA-N
SMILESCC(C)(C)C(N)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale brown
FormCrystals or powder or crystalline powder or fused solid or flakes
Melting Point (clear melt)151.0-157.0?C
Assay (GC)≥98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only