Thermo Scientific Chemicals

Potassium hydrogen phosphate, dried, 98+%, Thermo Scientific Chemicals

Catalog number: A11321.30
250 g, Each
Thermo Scientific Chemicals

Potassium hydrogen phosphate, dried, 98+%, Thermo Scientific Chemicals

Catalog number: A11321.30
250 g, Each
Quantity
Catalog number: A11321.30
also known as A11321-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7758-11-4
IUPAC Name
dipotassium hydrogen phosphate
Molecular Formula
HK2O4P
InChI Key
ZPWVASYFFYYZEW-UHFFFAOYSA-L
SMILES
[K+].[K+].OP([O-])([O-])=O
Appearance (Color)
White
Form
Powder or crystalline powder or granules
Assay (Non-aqueous acid-base Titration)
≥98.0 to ≤102.0% (dry wt. basis, non-U.S. specification)
Identification (FTIR)
Conforms (non-U.S. specification)
Assay from Suppliers CofA
≥98.0% (Titration, U.S. specification)

Description

Potassium hydrogen phosphate is used as a fertilizer, food additive, buffering agent, in molecular biology, biochemistry and chromatography. It is useful to increase the partition coefficients of amino acids in aqueous two-phase systems. It plays an important role for increase in rice grain yield by spraying chemicals including ferric chloride, dipotassium hydrogen phosphate and salicylic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium hydrogen phosphate is used as a fertilizer, food additive, buffering agent, in molecular biology, biochemistry and chromatography. It is useful to increase the partition coefficients of amino acids in aqueous two-phase systems. It plays an important role for increase in rice grain yield by spraying chemicals including ferric chloride, dipotassium hydrogen phosphate and salicylic acid.

Solubility
Soluble in water. Slightly soluble in alcohol.

Notes
Hygroscopic chemical and incompatible with strong oxidizing agents and acids. Keep the material away from heat, sparks and flame and sources of ignition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text