Thermo Scientific Chemicals

3,5-Bis(trifluoromethyl)benzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: A11325.14
25 g, Each
Thermo Scientific Chemicals

3,5-Bis(trifluoromethyl)benzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: A11325.14
25 g, Each
Quantity
Catalog number: A11325.14
also known as A11325-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
725-89-3
Appearance (Color)
White to pale cream
Melting Point (clear melt)
140.0-147.0?C
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Form
Crystals or powder or crystalline powder
Assay (Silylated GC)
≥97.5%

Description

In vitro metabolism studies on a series of 3,5-bis(trifluoromethyl)benzyl ethers have identified 3,5-bis(trifluoromethyl)benzoic acid as a significant metabolite possibly arising via oxidation of the benzylic position. The reactivity of the hydroxy- propyl units was demonstrated by the esterification of por- phyrazine derivatives with 3,5-bis(trifluoromethyl)benzoic acid in the presence of dicyclohexylcarbodiimide and toluene- p-sulfonic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
In vitro metabolism studies on a series of 3,5-bis(trifluoromethyl)benzyl ethers have identified 3,5-bis(trifluoromethyl)benzoic acid as a significant metabolite possibly arising via oxidation of the benzylic position. The reactivity of the hydroxy- propyl units was demonstrated by the esterification of por- phyrazine derivatives with 3,5-bis(trifluoromethyl)benzoic acid in the presence of dicyclohexylcarbodiimide and toluene- p-sulfonic acid.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text