Thermo Scientific Chemicals

Guanosine, 98%, Thermo Scientific Chemicals

Catalog number: A11328.22
100 g, Each
Thermo Scientific Chemicals

Guanosine, 98%, Thermo Scientific Chemicals

Catalog number: A11328.22
100 g, Each
Quantity
Catalog number: A11328.22
also known as A11328-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
118-00-3
IUPAC Name
2-amino-9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-6,9-dihydro-1H-purin-6-one
Molecular Formula
C10H13N5O5
InChI Key
NYHBQMYGNKIUIF-UUOKFMHZSA-N
SMILES
NC1=NC2=C(N=CN2[C@@H]2O[C@H](CO)[C@@H](O)[C@H]2O)C(=O)N1
Identification (FTIR)
Conforms
Water Content (Karl Fischer Titration)
≤6%
Appearance (Color)
White to pale cream
Assay (Non-aqueous acid-base Titration)
≥97.5 to ≤102.5% (dry wt. basis)
Optical Rotation
-66° to -74° (c=1.5 in 0.1N NaOH)

Description

Guanosine is used as a constituent of nucleic acids. It is used in metallic paints, simulated pearls,plastics,cosmetics industry etc,. It has been also used in pharmacokinetics as a prodrug. Guanosine is used in cell culture applications as a precursor of GMP.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Guanosine is used as a constituent of nucleic acids. It is used in metallic paints, simulated pearls,plastics,cosmetics industry etc,. It has been also used in pharmacokinetics as a prodrug. Guanosine is used in cell culture applications as a precursor of GMP.

Solubility
Soluble in Formic Acid:Water (1:1): 50 mg/mL. Insoluble in methanol, diethyl ether.

Notes
Store away from extreme heat. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text