1-Phenyl-1-butyne, 98%, Thermo Scientific Chemicals
1-Phenyl-1-butyne, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Phenyl-1-butyne, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
10 g
25 g
Catalog number A11332.06
also known as A11332-06
Price (USD)/ Each
36.40
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
36.40
Add to cart
1-Phenyl-1-butyne, 98%, Thermo Scientific Chemicals
Catalog numberA11332.06
Price (USD)/ Each
36.40
-
Add to cart
Chemical Identifiers
CAS622-76-4
IUPAC Name(but-1-yn-1-yl)benzene
Molecular FormulaC10H10
InChI KeyFFFMSANAQQVUJA-UHFFFAOYSA-N
SMILESCCC#CC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥97.5%
Refractive Index1.5500-1.5540 @ 20?C
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only