Thermo Scientific Chemicals

3,5-Dimethylphenylhydrazine hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: A11336.14
25 g, Each
Thermo Scientific Chemicals

3,5-Dimethylphenylhydrazine hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: A11336.14
25 g, Each
Quantity
Catalog number: A11336.14
also known as A11336-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
60481-36-9
IUPAC Name
(3,5-dimethylphenyl)hydrazine hydrochloride
Molecular Formula
C8H13ClN2
InChI Key
RSBQANBNDXZFIY-UHFFFAOYSA-N
SMILES
Cl.CC1=CC(NN)=CC(C)=C1
Appearance (Color)
White to cream to pale brown
Assay (Titration ex Chloride)
≥96.0 to ≤104.0%
Form
Powder or lumps

Description

3,5-Dimethylphenylhydrazine hydrochloride participates inFischer indole synthesis protocol in biological evaluation of novel antitubercular indolecarboxamide derivatives against drug-susceptible and drug-resistant mycobacterium tuberculosis strains.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dimethylphenylhydrazine hydrochloride participates inFischer indole synthesis protocol in biological evaluation of novel antitubercular indolecarboxamide derivatives against drug-susceptible and drug-resistant mycobacterium tuberculosis strains.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text