1-Bromodecane is used to produce decanal. It is also used in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Bromodecane is used to produce decanal. It is also used in organic synthesis.
Solubility
Miscible with most common organic solvents like ethanol, ethyl ether and chloroform. Not miscible in water.
Notes
Do not store together with strongly basic or oxidizing materials. Incompatible with strong oxidizing agents, strong bases.
RUO – Research Use Only
Gary M. McNamee; Brian C. Willett; Daniel M. La Perriere; Dennis G. Peters. Electrochemical reduction of 1-iododecane and 1-bromodecane at a mercury cathode in dimethylformamide. J. Am. Chem. Soc.1977, 99, (6)1831-1835.