Thermo Scientific Chemicals

1-Bromodecane, 98%, Thermo Scientific Chemicals

Catalog number: A11340.0E
2500 g, Each
Thermo Scientific Chemicals

1-Bromodecane, 98%, Thermo Scientific Chemicals

Catalog number: A11340.0E
2500 g, Each
Quantity
Catalog number: A11340.0E
also known as A11340-0E
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
112-29-8
IUPAC Name
1-bromodecane
Molecular Formula
C10H21Br
InChI Key
MYMSJFSOOQERIO-UHFFFAOYSA-N
SMILES
CCCCCCCCCCBr
Appearance (Color)
Clear colorless to white to pale yellow
Form
Liquid
Identification (FTIR)
Conforms
Assay (GC)
≥97.5%
Refractive Index
1.4540-1.4580 @ 20?C

Description

1-Bromodecane is used to produce decanal. It is also used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromodecane is used to produce decanal. It is also used in organic synthesis.

Solubility
Miscible with most common organic solvents like ethanol, ethyl ether and chloroform. Not miscible in water.

Notes
Do not store together with strongly basic or oxidizing materials. Incompatible with strong oxidizing agents, strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text