Thermo Scientific Chemicals

Pentadecanolide, 98%, Thermo Scientific Chemicals

Catalog number: A11341.14
25 g, Each
Thermo Scientific Chemicals

Pentadecanolide, 98%, Thermo Scientific Chemicals

Catalog number: A11341.14
25 g, Each
Quantity
Catalog number: A11341.14
also known as A11341-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
106-02-5
Assay (GC)
≥97.5%
Form
Fused solid
Melting Point (clear melt)
32.0-38.0?C
Appearance (Color)
White

Description

Pentadecanolide is a monomer used to make PGA-co-pentadecalactone polymers to be used in the development of directly compressed prolonged release tablets of slightly soluble drugs such as ketoprofen. Pentadecanolide may be used for the synthesis of poly(pentadecanolide) (PPDL) by Novozyme 435 catalyzed ring-opening polymerization. It may be employed as starting reagent for the synthesis of terminally-branched iso-fatty acids (iso-C15-C17).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pentadecanolide is a monomer used to make PGA-co-pentadecalactone polymers to be used in the development of directly compressed prolonged release tablets of slightly soluble drugs such as ketoprofen. Pentadecanolide may be used for the synthesis of poly(pentadecanolide) (PPDL) by Novozyme 435 catalyzed ring-opening polymerization. It may be employed as starting reagent for the synthesis of terminally-branched iso-fatty acids (iso-C15-C17).

Solubility
Soluble in alcohol, dipropylene glycol. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text