Thermo Scientific Chemicals

2,6-Dimethyl-p-benzoquinone, 99%, Thermo Scientific Chemicals

Catalog number: A11342.03
1 g, Each
Thermo Scientific Chemicals

2,6-Dimethyl-p-benzoquinone, 99%, Thermo Scientific Chemicals

Catalog number: A11342.03
1 g, Each
Quantity
Catalog number: A11342.03
also known as A11342-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
527-61-7
IUPAC Name
2,6-dimethylcyclohexa-2,5-diene-1,4-dione
Molecular Formula
C8H8O2
InChI Key
SENUUPBBLQWHMF-UHFFFAOYSA-N
SMILES
CC1=CC(=O)C=C(C)C1=O
Appearance (Color)
Yellow to orange to brown
Melting Point (clear melt)
67.0-74.0?C
Assay (GC)
≥98.5%
Form
Crystals or powder or crystalline powder

Description

2,6-Dimethyl-p-benzoquinone is used as organic intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dimethyl-p-benzoquinone is used as organic intermediates.

Solubility
Soluble in chloroform. Insoluble in water.

Notes
Incompatible with strong oxidizing agents and reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text