Thermo Scientific Chemicals

3,4-Dimethoxyphenol, 98%, Thermo Scientific Chemicals

Catalog number: A11344.14
25 g, Each
Thermo Scientific Chemicals

3,4-Dimethoxyphenol, 98%, Thermo Scientific Chemicals

Catalog number: A11344.14
25 g, Each
Quantity
Catalog number: A11344.14
also known as A11344-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2033-89-8
IUPAC Name
3,4-dimethoxyphenol
Molecular Formula
C8H10O3
InChI Key
SMFFZOQLHYIRDA-UHFFFAOYSA-N
SMILES
COC1=CC=C(O)C=C1OC
Form
Powder or crystalline powder
Identification (FTIR)
Conforms
Assay (GC)
≥97.5%
Appearance (Color)
White to cream to brown or pale pink
Comment
May darken on storage

Description

3,4-Dimethoxyphenol is used in the preparation of 5,6-dimethoxy benzofuranone derivatives and multi-target anti Alzheimer compounds. It is also used in the preparation of 3,4-dimethoxyphenyl-beta-D-glucopyranoside and 4-(but-2-enyloxy)-1,2-dimethoxybenzene. Further, it serves as a precursor for 4H-chromenes synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Dimethoxyphenol is used in the preparation of 5,6-dimethoxy benzofuranone derivatives and multi-target anti Alzheimer compounds. It is also used in the preparation of 3,4-dimethoxyphenyl-beta-D-glucopyranoside and 4-(but-2-enyloxy)-1,2-dimethoxybenzene. Further, it serves as a precursor for 4H-chromenes synthesis.

Solubility
Soluble in water.

Notes
Light sensitive. Store in a cool place. Incompatible with bases, acid chlorides, acid anhydrides and oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text