Thermo Scientific Chemicals

5-Chlorothiophene-2-sulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: A11348.09
10 g, Each
Thermo Scientific Chemicals

5-Chlorothiophene-2-sulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: A11348.09
10 g, Each
Quantity
Catalog number: A11348.09
also known as A11348-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2766-74-7
IUPAC Name
5-chlorothiophene-2-sulfonyl chloride
Molecular Formula
C4H2Cl2O2S2
InChI Key
SORSTNOXGOXWAO-UHFFFAOYSA-N
SMILES
ClC1=CC=C(S1)S(Cl)(=O)=O
Refractive Index
1.5855-1.5905 @ 20?C
Form
Liquid
Appearance (Color)
Clear colorless to pale yellow to yellow to brown
Assay (GC)
≥96.0%

Description

5-Chlorothiophene-2-sulfonyl chloride is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs. It is used in the preparation of Notch-1-sparing γ-secretase inhibitors for the treatment of Alzheimer's disease.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Not miscible in water.

Notes
Moisture Sensitive. Store at 4°C. Incomaptible with oxidizing agents and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text