Thermo Scientific Chemicals

D-Serine, 99%

Catalog number: A11353.22
100 g, Each
Thermo Scientific Chemicals

D-Serine, 99%

Catalog number: A11353.22
100 g, Each
Quantity
Catalog number: A11353.22
also known as A11353-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
312-84-5
IUPAC Name
2-amino-3-hydroxypropanoic acid
Molecular Formula
C3H7NO3
InChI Key
MTCFGRXMJLQNBG-UHFFFAOYNA-N
SMILES
NC(CO)C(O)=O
Optical Rotation
-13.4 ± 0.3? (C=5 in 5N HCl)
Form
Crystals or powder or crystalline powder
Appearance (Color)
White to pale cream
Assay (Non-aqueous acid-base Titration)
≥98.5 to ≤101.5%

Description

D-Serine is involved in the biosynthesis of purines, pyrimidines, and other amino acids. D-Serine also is an agonist of glycine site of the NMDA-type glutamate receptor. It also acts as Lacosamide intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Serine is involved in the biosynthesis of purines, pyrimidines, and other amino acids. D-Serine also is an agonist of glycine site of the NMDA-type glutamate receptor. It also acts as Lacosamide intermediate.

Solubility
Soluble in water (346 mg/ml at 20°C), and mineral acids. Insoluble in alcohol, ether, and benzene.

Notes
Store at 4°C. Store away from oxidizing agents. Keep container closed when not in use.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text