Thermo Scientific Chemicals

1,3-Bis(trifluoromethyl)benzene, 98+%, Thermo Scientific Chemicals

Catalog number: A11360.18
50 g, Each
Thermo Scientific Chemicals

1,3-Bis(trifluoromethyl)benzene, 98+%, Thermo Scientific Chemicals

Catalog number: A11360.18
50 g, Each
Quantity
Catalog number: A11360.18
also known as A11360-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
402-31-3
IUPAC Name
1,3-bis(trifluoromethyl)benzene
Molecular Formula
C8H4F6
InChI Key
SJBBXFLOLUTGCW-UHFFFAOYSA-N
SMILES
FC(F)(F)C1=CC(=CC=C1)C(F)(F)F
Form
Liquid
Refractive Index
1.3780-1.3820 @ 20?C
Appearance (Color)
Clear colorless
Identification (FTIR)
Conforms
Assay (GC)
≥98.0%

Description

1,3-Bis(trifluoromethyl)benzene undergoes regioselective metalation and subsequent carboxylation at position 2 to give 2,6-bis(trifluoromethyl)benzoic acid. a convenient, selective synthesis of bis[2,4-bis(trifluoromethyl)phenyl]phosphane derivatives with 1,3-bis(trifluoromethyl)benzene as starting material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Bis(trifluoromethyl)benzene undergoes regioselective metalation and subsequent carboxylation at position 2 to give 2,6-bis(trifluoromethyl)benzoic acid. a convenient, selective synthesis of bis[2,4-bis(trifluoromethyl)phenyl]phosphane derivatives with 1,3-bis(trifluoromethyl)benzene as starting material.

Solubility
Insoluble in water. Soluble in alcohol, ether, benzene.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text