Thermo Scientific Chemicals

Ethyl 4-bromobenzoate, 98+%, Thermo Scientific Chemicals

Catalog number: A11365.09
10 g, Each
Thermo Scientific Chemicals

Ethyl 4-bromobenzoate, 98+%, Thermo Scientific Chemicals

Catalog number: A11365.09
10 g, Each
Quantity
Catalog number: A11365.09
also known as A11365-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5798-75-4
IUPAC Name
ethyl 4-bromobenzoate
Molecular Formula
C9H9BrO2
InChI Key
XZIAFENWXIQIKR-UHFFFAOYSA-N
SMILES
CCOC(=O)C1=CC=C(Br)C=C1
Form
Liquid
Refractive Index
1.5440-1.5480 @ 20?C
Appearance (Color)
Clear colorless to yellow
Assay (GC)
≥98.0%

Description

Ethyl 4-bromobenzoate undergoes reduction with potassium diisobutyl-t-butoxyaluminum hydride (PDBBA) at 0°C to yield aldehydes. Reaction of ethyl 4-bromobenzoate and substituted benzyl chloride with zinc dust and a Pd catalyst is reported.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 4-bromobenzoate undergoes reduction with potassium diisobutyl-t-butoxyaluminum hydride (PDBBA) at 0°C to yield aldehydes. Reaction of ethyl 4-bromobenzoate and substituted benzyl chloride with zinc dust and a Pd catalyst is reported.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text