1-Hexadecyne, tech. 90%, Thermo Scientific Chemicals
1-Hexadecyne, tech. 90%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Hexadecyne, tech. 90%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number A11366.14
also known as A11366-14
Price (USD)/ Each
440.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
440.00
Add to cart
1-Hexadecyne, tech. 90%, Thermo Scientific Chemicals
Catalog numberA11366.14
Price (USD)/ Each
440.00
-
Add to cart
Chemical Identifiers
CAS629-74-3
SpecificationsSpecification SheetSpecification Sheet
Formliquid
Refractive Index1.4380 - 1.4460 @20?C
Appearance (Color)Clear, colourless to yellow
Assay (GC)> 88.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only