Thermo Scientific Chemicals

Tetra-n-hexylammonium bromide, 98%, Thermo Scientific Chemicals

Catalog number: A11372.18
50 g, Each
Thermo Scientific Chemicals

Tetra-n-hexylammonium bromide, 98%, Thermo Scientific Chemicals

Catalog number: A11372.18
50 g, Each
Quantity
Catalog number: A11372.18
also known as A11372-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4328-13-6
IUPAC Name
tetrahexylazanium bromide
Molecular Formula
C24H52BrN
InChI Key
SYZCZDCAEVUSPM-UHFFFAOYSA-M
SMILES
[Br-].CCCCCC[N+](CCCCCC)(CCCCCC)CCCCCC
Appearance (Color)
White to cream or pale yellow
Form
Powder or crystalline powder or flakes
Assay (Titration ex Bromide)
≥97.5 to ≤102.5%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text