Thermo Scientific Chemicals

Amido Black 10B, Thermo Scientific Chemicals

Catalog number: A11374.14
25 g, Each
Thermo Scientific Chemicals

Amido Black 10B, Thermo Scientific Chemicals

Catalog number: A11374.14
25 g, Each
Quantity
Catalog number: A11374.14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1064-48-8
IUPAC Name
disodium (3Z)-5-amino-6-[2-(4-nitrophenyl)diazen-1-yl]-4-oxo-3-(2-phenylhydrazin-1-ylidene)-3,4-dihydronaphthalene-2,7-disulfonate
Molecular Formula
C22H14N6Na2O9S2
InChI Key
HKBVRFLHNUEVRO-DWTBGCDMSA-L
SMILES
[Na+].[Na+].NC1=C(N=NC2=CC=C(C=C2)[N+]([O-])=O)C(=CC2=C1C(=O)\C(=N\NC1=CC=CC=C1)C(=C2)S([O-])(=O)=O)S([O-])(=O)=O
Appearance (Color)
Brown to black
Form
Powder
Loss on Drying
≤10% (105°C, constant weight)
Solution Test
(1mg/L in water): Clear, dark to very dark blue solution
Specific Absorbance (E 1%/1cm)
≥450 at lambda-max (non-U.S. sourced material)

Description

Amido Black 10B is used as a histological, protein, and nucleic acid dye used in electrophoresis. Naphthol Blue Black is an important and commonly used industrial dye. This dye is also used in rapid staining of protein bands on nitrocellulose membranes such as Western blots. Naphthol Blue Black can be utilized as a nucleic acid stain in SDS-Page gels. Additionally this compound has been untilized in cell proliferation assays and histochemical studies. It is also used in criminal investigations to detect blood present with latent fingerprints. They are also used in paints, inks, plastics and leather.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text