Thermo Scientific Chemicals

Tris(hydroxymethyl)aminomethane hydrochloride, 99+%

Catalog number: A11379.30
250 g, Each
Thermo Scientific Chemicals

Tris(hydroxymethyl)aminomethane hydrochloride, 99+%

Catalog number: A11379.30
250 g, Each
Quantity
Catalog number: A11379.30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1185-53-1
IUPAC Name
hydrogen 2-amino-2-(hydroxymethyl)propane-1,3-diol chloride
Molecular Formula
C4H12ClNO3
InChI Key
QKNYBSVHEMOAJP-UHFFFAOYSA-N
SMILES
[H+].[Cl-].NC(CO)(CO)CO
Appearance (Color)
White
Assay (Non-aqueous acid-base Titration)
≥99.0% to ≤101.0%
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms

Description

Tris(hydroxymethyl)aminomethane hydrochloride is used as a component of buffer solutions in biology, biochemistry and molecular biology. It is also used for crystallization of proteins at various pH values. Further, it inhibits motor responses to adrenergic motor nerve stimulation in smooth muscles. In addition to this, it is used in coelomic fluid and in cortland medium to store unfertilized rainbow trout eggs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tris(hydroxymethyl)aminomethane hydrochloride is used as a component of buffer solutions in biology, biochemistry and molecular biology. It is also used for crystallization of proteins at various pH values. Further, it inhibits motor responses to adrenergic motor nerve stimulation in smooth muscles. In addition to this, it is used in coelomic fluid and in cortland medium to store unfertilized rainbow trout eggs.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents, strong acids and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text